Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fastkd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177244.3 | NP_796218.2 | 5 - 6 | 1190 | 179 | |
GenBank mRNA | AK220423.1 | - | 1 - 2 | 125 | 179 | |
BC034292.1 | - | 2 - 3 | 297 | 179 | ||
BC069943.1 | - | 4 - 5 | 683 | 179 | ||
BC075689.1 | - | 5 - 6 | 1190 | 30609755 | 179 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |