Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tlk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172664.3 | NP_766252.2 | 12 - 13 | 1390 | 100 | |
GenBank mRNA | AK029773.1 | - | 11 - 12 | 1116 | 100 | |
AK172897.1 | - | 6 - 7 | 654 | 100 | ||
BC033931.1 | - | 10 - 11 | 952 | 100 | ||
BC051641.1 | - | 12 - 13 | 1390 | 6507968 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |