Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sco1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040026.1 | NP_001035115.1 | 2 - 3 | 335 | 76 | |
GenBank mRNA | AK017794.1 | - | 2 - 3 | 336 | 76 | |
AK139512.1 | - | 2 - 3 | 306 | 76 | ||
AK167305.1 | - | 2 - 3 | 200 | 76 | ||
BC139008.1 | - | 2 - 3 | 330 | 8862030 | 76 | |
BC139009.1 | - | 2 - 3 | 330 | 8862031 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |