Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100007167.mm.1 |
NM_001013013.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007167.mm.1 |
NM_001013013.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007167.mm.1 |
TC1100000864.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100007159.mm.1 |
NM_001013013.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007159.mm.1 |
NM_001013013.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007159.mm.1 |
TC1100000864.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100007161.mm.1 |
NM_001013013.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007161.mm.1 |
NM_001013013.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007161.mm.1 |
TC1100000864.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100007160.mm.1 |
NM_001013013.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007160.mm.1 |
NM_001013013.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007160.mm.1 |
TC1100000864.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100007168.mm.1 |
NM_001013013.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007168.mm.1 |
NM_001013013.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007168.mm.1 |
TC1100000864.mm.1 |
MTA TC |