Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100007002.mm.1 |
NM_177369.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007002.mm.1 |
NM_177369.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007002.mm.1 |
TC1100000856.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100007004.mm.1 |
NM_177369.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007004.mm.1 |
NM_177369.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007004.mm.1 |
TC1100000856.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100006990.mm.1 |
NM_177369.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006990.mm.1 |
NM_177369.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006990.mm.1 |
TC1100000856.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100006992.mm.1 |
NM_177369.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006992.mm.1 |
NM_177369.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006992.mm.1 |
TC1100000856.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100006996.mm.1 |
NM_177369.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006996.mm.1 |
NM_177369.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006996.mm.1 |
TC1100000856.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100006994.mm.1 |
NM_177369.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006994.mm.1 |
NM_177369.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006994.mm.1 |
TC1100000856.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100007000.mm.1 |
NM_177369.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007000.mm.1 |
NM_177369.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007000.mm.1 |
TC1100000856.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100007005.mm.1 |
NM_177369.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007005.mm.1 |
NM_177369.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007005.mm.1 |
TC1100000856.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100007006.mm.1 |
NM_177369.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007006.mm.1 |
NM_177369.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007006.mm.1 |
TC1100000856.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100006989.mm.1 |
NM_177369.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006989.mm.1 |
NM_177369.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006989.mm.1 |
TC1100000856.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100007001.mm.1 |
NM_177369.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007001.mm.1 |
NM_177369.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007001.mm.1 |
TC1100000856.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100006995.mm.1 |
NM_177369.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006995.mm.1 |
NM_177369.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006995.mm.1 |
TC1100000856.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100006997.mm.1 |
NM_177369.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006997.mm.1 |
NM_177369.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006997.mm.1 |
TC1100000856.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100007003.mm.1 |
NM_177369.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007003.mm.1 |
NM_177369.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007003.mm.1 |
TC1100000856.mm.1 |
MTA TC |