Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gfra4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136063.2 | NP_001129535.1 | 2 - 3 | 407 | 91 | |
NM_001271001.1 | NP_001257930.1 | 2 - 3 | 407 | 91 | ||
NM_001271002.1 | NP_001257931.1 | 2 - 3 | 434 | 91 | ||
NM_020014.2 | NP_064398.1 | 2 - 3 | 434 | 91 | ||
GenBank mRNA | AJ276514.1 | - | 2 - 3 | 434 | 91 | |
AJ276516.1 | - | 2 - 3 | 434 | 91 | ||
AJ276870.1 | - | 2 - 3 | 407 | 91 | ||
AJ276871.1 | - | 2 - 3 | 407 | 91 | ||
BC111808.1 | - | 2 - 3 | 433 | 91 | ||
BC141873.1 | - | 2 - 3 | 429 | 91 | ||
BC141874.1 | - | 2 - 3 | 434 | 40040202 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |