Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200039691.mm.1 |
NM_001081162.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200039691.mm.1 |
NM_001081162.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200039691.mm.1 |
TC0200004636.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200039694.mm.1 |
NM_001081162.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200039694.mm.1 |
NM_001081162.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200039694.mm.1 |
TC0200004636.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200039692.mm.1 |
NM_001081162.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200039692.mm.1 |
NM_001081162.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200039692.mm.1 |
TC0200004636.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200039693.mm.1 |
NM_001081162.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200039693.mm.1 |
NM_001081162.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200039693.mm.1 |
TC0200004636.mm.1 |
MTA TC |