Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspa12b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028306.3 | NP_082582.1 | 2 - 3 | 165 | 117 | |
GenBank mRNA | AK012548.1 | - | 2 - 3 | 138 | 117 | |
AY196790.1 | - | 1 - 2 | 45 | 117 | ||
BC011103.1 | - | 2 - 3 | 165 | 3990088 | 117 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |