Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163529.1 | NP_001157001.1 | 14 - 15 | 1900 | 71 | |
NM_033615.2 | NP_291093.2 | 14 - 15 | 1900 | 71 | ||
GenBank mRNA | AB059632.1 | - | 14 - 15 | 1900 | 71 | |
AB059633.1 | - | 14 - 15 | 1900 | 71 | ||
AF386072.1 | - | 10 - 11 | 1267 | 71 | ||
AF472524.1 | - | 14 - 15 | 1632 | 71 | ||
AK048367.1 | - | 10 - 11 | 1167 | 71 | ||
AY054984.1 | - | 14 - 15 | 1600 | 71 | ||
AY382193.1 | - | 14 - 15 | 1631 | 71 | ||
AY382194.1 | - | 14 - 15 | 1631 | 71 | ||
AY953137.1 | - | 9 - 10 | 1175 | 71 | ||
BC138139.1 | - | 14 - 15 | 1623 | 8861158 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |