Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163529.1 | NP_001157001.1 | 15 - 16 | 2003 | 61 | |
NM_033615.2 | NP_291093.2 | 15 - 16 | 2003 | 61 | ||
GenBank mRNA | AB059632.1 | - | 15 - 16 | 2003 | 61 | |
AB059633.1 | - | 15 - 16 | 2003 | 61 | ||
AF386072.1 | - | 11 - 12 | 1370 | 61 | ||
AF472524.1 | - | 15 - 16 | 1735 | 61 | ||
AK048367.1 | - | 11 - 12 | 1270 | 61 | ||
AK053825.1 | - | 13 - 14 | 2261 | 61 | ||
AK138517.1 | - | 1 - 2 | 347 | 61 | ||
AY054984.1 | - | 15 - 16 | 1703 | 61 | ||
AY382193.1 | - | 15 - 16 | 1734 | 61 | ||
AY382194.1 | - | 15 - 16 | 1734 | 61 | ||
AY953137.1 | - | 10 - 11 | 1278 | 61 | ||
BC138139.1 | - | 15 - 16 | 1726 | 8861158 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |