Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025936.3 | NP_080212.2 | 8 - 9 | 1001 | 153 | |
GenBank mRNA | AK011383.1 | - | 8 - 9 | 979 | 153 | |
AK076160.1 | - | 8 - 9 | 976 | 153 | ||
AK149856.1 | - | 8 - 9 | 976 | 153 | ||
AK151168.1 | - | 2 - 3 | 167 | 153 | ||
AK151966.1 | - | 7 - 8 | 799 | 153 | ||
AK152108.1 | - | 8 - 9 | 976 | 153 | ||
AK160216.1 | - | 8 - 9 | 976 | 153 | ||
AK166097.1 | - | 8 - 9 | 976 | 153 | ||
AK168194.1 | - | 8 - 9 | 976 | 153 | ||
BC020132.1 | - | 8 - 9 | 959 | 3989952 | 153 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |