Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159502.1 | NP_001152974.1 | 6 - 7 | 2246 | 86 | |
NM_020494.3 | NP_065240.2 | 6 - 7 | 2080 | 86 | ||
GenBank mRNA | AF214732.1 | - | 6 - 7 | 2060 | 86 | |
AK149810.1 | - | 6 - 7 | 2033 | 86 | ||
AK150778.1 | - | 6 - 7 | 2032 | 86 | ||
AK152551.1 | - | 6 - 7 | 2032 | 86 | ||
AK152928.1 | - | 6 - 7 | 2032 | 86 | ||
AK166360.1 | - | 6 - 7 | 2041 | 86 | ||
AK172557.1 | - | 6 - 7 | 2033 | 86 | ||
BC043721.1 | - | 6 - 7 | 2048 | 86 | ||
BC055048.1 | - | 6 - 7 | 2010 | 6484820 | 86 | |
BC055317.1 | - | 6 - 7 | 2246 | 5696084 | 86 | |
U46690.1 | - | 4 - 5 | 794 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |