Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arl5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK164977.1 | - | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon |