Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200037930.mm.1 |
NM_178795.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037930.mm.1 |
NM_178795.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037930.mm.1 |
TC0200004485.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200037931.mm.1 |
NM_178795.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037931.mm.1 |
NM_178795.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037931.mm.1 |
TC0200004485.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200037933.mm.1 |
NM_178795.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037933.mm.1 |
NM_178795.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037933.mm.1 |
TC0200004485.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200037928.mm.1 |
NM_178795.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037928.mm.1 |
NM_178795.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037928.mm.1 |
TC0200004485.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200037929.mm.1 |
NM_178795.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037929.mm.1 |
NM_178795.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037929.mm.1 |
TC0200004485.mm.1 |
MTA TC |