Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lcn9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029959.2 | NP_084235.1 | 4 - 5 | 429 | 81 | |
GenBank mRNA | AF435738.1 | - | 4 - 5 | 429 | 81 | |
AK020305.1 | - | 4 - 5 | 431 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |