Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Qsox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK140324.1 | - | 3 - 4 | 497 | 104 | |
AK157992.1 | - | 3 - 4 | 490 | 104 | ||
AK165642.1 | - | 3 - 4 | 499 | 104 | ||
BC030934.1 | - | 3 - 4 | 531 | 4920915 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |