Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mamdc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081199.1 | NP_001074668.2 | 8 - 9 | 1046 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |