Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp6r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172894.2 | NP_766482.2 | 10 - 11 | 1767 | 125 | |
GenBank mRNA | BC053076.1 | - | 10 - 11 | 1767 | 5698212 | 125 |
BC141039.1 | - | 9 - 10 | 1314 | 9055651 | 125 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |