Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK051552.1 | - | 1 - 2 | 295 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |