Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277903.1 | NP_001264832.1 | 10 - 11 | 558 | 84 | |
NM_001277904.1 | NP_001264833.1 | 9 - 10 | 522 | 84 | ||
NM_011618.2 | NP_035748.1 | 10 - 11 | 555 | 84 | ||
GenBank mRNA | AF020946.1 | - | 10 - 11 | 542 | 84 | |
AJ131711.1 | - | 9 - 10 | 503 | 84 | ||
AK003317.1 | - | 2 - 3 | 137 | 84 | ||
AK160226.1 | - | 10 - 11 | 556 | 84 | ||
BC138871.1 | - | 10 - 11 | 535 | 8861893 | 84 | |
BC141142.1 | - | 10 - 11 | 538 | 9055754 | 84 | |
BC145449.1 | - | 9 - 10 | 502 | 9054007 | 84 | |
BC145450.1 | - | 9 - 10 | 502 | 9054008 | 84 | |
BC145451.1 | - | 10 - 11 | 538 | 9054009 | 84 | |
U92883.1 | - | 10 - 11 | 539 | 84 | ||
U92884.1 | - | 9 - 10 | 506 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |