Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efcab11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030172.2 | NP_084448.1 | 2 - 3 | 257 | 89 | |
GenBank mRNA | AK011507.1 | - | 2 - 3 | 257 | 89 | |
AK011836.1 | - | 2 - 3 | 239 | 89 | ||
AK035051.1 | - | 2 - 3 | 237 | 89 | ||
BC049109.1 | - | 2 - 3 | 219 | 6414718 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |