Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cts8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019541.3 | NP_062414.3 | 5 - 6 | 706 | 65 | |
GenBank mRNA | AF250840.1 | - | 5 - 6 | 718 | 65 | |
AK049474.1 | - | 5 - 6 | 707 | 65 | ||
AY014780.1 | - | 5 - 6 | 705 | 65 | ||
BC068241.1 | - | 5 - 6 | 696 | 30019120 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |