Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtpbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027000.4 | NP_081276.2 | 7 - 8 | 856 | 152 | |
GenBank mRNA | AF348208.1 | - | 7 - 8 | 863 | 152 | |
AK011580.1 | - | 7 - 8 | 867 | 152 | ||
AK141852.1 | - | 7 - 8 | 866 | 152 | ||
AK164330.1 | - | 7 - 8 | 857 | 152 | ||
BC005514.1 | - | 7 - 8 | 837 | 152 | ||
BC025431.1 | - | 7 - 8 | 861 | 5007174 | 152 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |