Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adarb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_052977.5 | NP_443209.2 | 9 - 10 | 2417 | 74 | |
GenBank mRNA | AF232942.1 | - | 9 - 10 | 2034 | 74 | |
AF270495.1 | - | 9 - 10 | 2134 | 74 | ||
AK033916.1 | - | 9 - 10 | 2356 | 74 | ||
AK045538.1 | - | 9 - 10 | 2357 | 74 | ||
AK134964.1 | - | 9 - 10 | 2419 | 74 | ||
AK139079.1 | - | 9 - 10 | 2375 | 74 | ||
AK139273.1 | - | 9 - 10 | 2375 | 74 | ||
BC052426.1 | - | 9 - 10 | 2342 | 6835942 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |