Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myh4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010855.3 | NP_034985.2 | 20 - 21 | 2402 | 69 | |
GenBank mRNA | AK138608.1 | - | 20 - 21 | 2403 | 69 | |
BC006031.1 | - | 5 - 6 | 473 | 69 | ||
BC141362.1 | - | 18 - 19 | 2319 | 9055974 | 69 | |
CX730791.1 | - | 5 - 6 | 493 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons |