Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100006866.mm.1 |
NM_001039545.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006866.mm.1 |
NM_001039545.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006866.mm.1 |
TC1100000853.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100006868.mm.1 |
NM_001039545.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006868.mm.1 |
NM_001039545.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006868.mm.1 |
TC1100000853.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100006864.mm.1 |
NM_001039545.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006864.mm.1 |
NM_001039545.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006864.mm.1 |
TC1100000853.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100006869.mm.1 |
NM_001039545.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006869.mm.1 |
NM_001039545.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006869.mm.1 |
TC1100000853.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100006865.mm.1 |
NM_001039545.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006865.mm.1 |
NM_001039545.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006865.mm.1 |
TC1100000853.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100006867.mm.1 |
NM_001039545.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006867.mm.1 |
NM_001039545.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006867.mm.1 |
TC1100000853.mm.1 |
MTA TC |