Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdm6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017426.1 | NP_001017426.1 | 12 - 13 | 3969 | 76 | |
GenBank mRNA | AK129121.1 | - | 2 - 3 | 758 | 76 | |
AK136019.1 | - | 2 - 3 | 150 | 76 | ||
BC075632.1 | - | 13 - 14 | 4145 | 30362899 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |