Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dclre1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110214.1 | NP_001103684.1 | 10 - 11 | 1001 | 85 | |
NM_146114.3 | NP_666226.2 | 10 - 11 | 1001 | 85 | ||
NM_175683.4 | NP_783614.1 | 10 - 11 | 1001 | 85 | ||
GenBank mRNA | AF387731.1 | - | 10 - 11 | 983 | 85 | |
AK037126.1 | - | 10 - 11 | 968 | 85 | ||
AK052369.1 | - | 10 - 11 | 957 | 85 | ||
AK052490.1 | - | 10 - 11 | 940 | 85 | ||
AK088810.1 | - | 8 - 9 | 783 | 85 | ||
AK166324.1 | - | 10 - 11 | 944 | 85 | ||
BC108935.2 | - | 10 - 11 | 954 | 40039591 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |