Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mok | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011973.2 | NP_036103.1 | 5 - 6 | 547 | 112 | |
GenBank mRNA | AB022695.1 | - | 5 - 6 | 547 | 112 | |
BC049604.1 | - | 3 - 4 | 374 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |