Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clmn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040682.1 | NP_001035772.1 | 10 - 11 | 2887 | 62 | |
NM_053155.2 | NP_444385.2 | 10 - 11 | 2887 | 62 | ||
NR_104439.1 | 10 - 11 | 2887 | 62 | |||
GenBank mRNA | AB047978.1 | - | 10 - 11 | 2880 | 62 | |
AB059643.1 | - | 10 - 11 | 2880 | 62 | ||
AB059644.1 | - | 10 - 11 | 2880 | 62 | ||
AK077023.1 | - | 7 - 8 | 2469 | 62 | ||
AK220261.1 | - | 2 - 3 | 866 | 62 | ||
BC141280.1 | - | 10 - 11 | 2820 | 9055892 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |