Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clmn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040682.1 | NP_001035772.1 | 2 - 3 | 269 | 103 | |
NM_053155.2 | NP_444385.2 | 2 - 3 | 269 | 103 | ||
NR_104435.1 | 2 - 3 | 269 | 103 | |||
NR_104436.1 | 2 - 3 | 269 | 103 | |||
NR_104437.1 | 2 - 3 | 269 | 103 | |||
NR_104438.1 | 2 - 3 | 269 | 103 | |||
NR_104439.1 | 2 - 3 | 269 | 103 | |||
GenBank mRNA | AB047978.1 | - | 2 - 3 | 262 | 103 | |
AB059643.1 | - | 2 - 3 | 262 | 103 | ||
AB059644.1 | - | 2 - 3 | 262 | 103 | ||
AB059645.1 | - | 2 - 3 | 262 | 103 | ||
AB059646.1 | - | 2 - 3 | 262 | 103 | ||
AB059647.1 | - | 2 - 3 | 262 | 103 | ||
AB059648.1 | - | 2 - 3 | 262 | 103 | ||
AK147777.1 | - | 2 - 3 | 166 | 103 | ||
BC019176.1 | - | 2 - 3 | 200 | 103 | ||
BC096470.1 | - | 2 - 3 | 158 | 103 | ||
BC141280.1 | - | 2 - 3 | 202 | 9055892 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |