Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nqo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020282.3 | NP_064678.1 | 1 - 2 | 142 | 74 | |
GenBank mRNA | BC027629.1 | - | 1 - 2 | 50 | 1225122 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |