Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nqo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163239.1 | NP_001156711.1 | 3 - 4 | 435 | 66 | |
NM_001163241.1 | NP_001156713.1 | 4 - 5 | 523 | 66 | ||
NM_001163242.1 | NP_001156714.1 | 4 - 5 | 452 | 66 | ||
NM_020282.3 | NP_064678.1 | 5 - 6 | 605 | 66 | ||
GenBank mRNA | AF252260.1 | - | 4 - 5 | 451 | 66 | |
AK008138.1 | - | 3 - 4 | 407 | 66 | ||
AK008465.1 | - | 3 - 4 | 410 | 66 | ||
AK010842.1 | - | 4 - 5 | 495 | 66 | ||
AK049781.1 | - | 4 - 5 | 461 | 66 | ||
BC027629.1 | - | 5 - 6 | 513 | 1225122 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |