Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gcnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133219.1 | NP_573482.1 | 1 - 2 | 1169 | 114 | |
GenBank mRNA | AK019924.1 | - | 1 - 2 | 1154 | 114 | |
AK146638.1 | - | 1 - 2 | 271 | 114 | ||
AK146679.1 | - | 1 - 2 | 271 | 114 | ||
AK169167.1 | - | 1 - 2 | 271 | 114 | ||
AY236875.1 | - | 1 - 2 | 1202 | 114 | ||
AY435150.1 | - | 1 - 2 | 920 | 114 | ||
BC054845.1 | - | 1 - 2 | 1151 | 6493137 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |