Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pygb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153781.1 | NP_722476.1 | 18 - 19 | 2381 | 73 | |
GenBank mRNA | AK032769.1 | - | 18 - 19 | 2394 | 73 | |
AK035673.1 | - | 18 - 19 | 2111 | 73 | ||
AK133794.1 | - | 9 - 10 | 1111 | 73 | ||
AK142716.1 | - | 18 - 19 | 2391 | 73 | ||
AK147763.1 | - | 18 - 19 | 2413 | 73 | ||
AK168381.1 | - | 18 - 19 | 2393 | 73 | ||
AK169052.1 | - | 18 - 19 | 2416 | 73 | ||
BC031457.1 | - | 18 - 19 | 2388 | 73 | ||
BC032209.1 | - | 12 - 13 | 1536 | 73 | ||
BC035283.1 | - | 18 - 19 | 2381 | 4947455 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |