Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146025.1 | NP_001139497.1 | 9 - 10 | 1499 | 68 | |
NM_001146026.1 | NP_001139498.1 | 9 - 10 | 1424 | 68 | ||
NM_001146027.1 | NP_001139499.1 | 9 - 10 | 1290 | 68 | ||
NM_134064.2 | NP_598825.2 | 9 - 10 | 1432 | 68 | ||
NR_027395.1 | 9 - 10 | 1352 | 68 | |||
NR_027396.1 | 9 - 10 | 1349 | 68 | |||
GenBank mRNA | AK147349.1 | - | 9 - 10 | 1425 | 68 | |
AK160090.1 | - | 9 - 10 | 1283 | 68 | ||
AK160115.1 | - | 9 - 10 | 1283 | 68 | ||
AK161651.1 | - | 9 - 10 | 1414 | 68 | ||
AK171943.1 | - | 9 - 10 | 1281 | 68 | ||
AK172257.1 | - | 9 - 10 | 1282 | 68 | ||
BC017630.1 | - | 9 - 10 | 1225 | 4507289 | 68 | |
BC035548.1 | - | 9 - 10 | 1247 | 4221284 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |