Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sephs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175400.6 | NP_780609.4 | 1 - 2 | 292 | 71 | |
GenBank mRNA | AK044971.1 | - | 1 - 2 | 229 | 71 | |
BC066037.1 | - | 1 - 2 | 291 | 6405661 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |