Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Optn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181848.4 | NP_862896.1 | 5 - 6 | 699 | 64 | |
GenBank mRNA | AY071834.1 | - | 5 - 6 | 806 | 64 | |
BC061185.1 | - | 5 - 6 | 699 | 30314331 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |