Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080554.2 | NP_542121.2 | 4 - 5 | 596 | 140 | |
GenBank mRNA | AK005335.1 | - | 4 - 5 | 574 | 140 | |
AK011026.1 | - | 4 - 5 | 573 | 140 | ||
AK029067.1 | - | 3 - 4 | 893 | 140 | ||
AK032992.1 | - | 4 - 5 | 574 | 140 | ||
AK078419.1 | - | 4 - 5 | 574 | 140 | ||
AK145587.1 | - | 4 - 5 | 573 | 140 | ||
AK148284.1 | - | 597 | 140 | |||
AK220321.1 | - | 4 - 5 | 537 | 140 | ||
BC019112.1 | - | 4 - 5 | 607 | 140 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |