Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppapdc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK048016.1 | - | 2 - 3 | 240 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |