Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Golga2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080968.1 | NP_001074437.1 | 20 - 21 | 2180 | 83 | |
NM_133852.2 | NP_598613.2 | 21 - 22 | 2261 | 83 | ||
GenBank mRNA | AK076370.1 | - | 15 - 16 | 1879 | 83 | |
AK220509.1 | - | 21 - 22 | 2189 | 83 | ||
BC011407.1 | - | 19 - 20 | 1981 | 3596079 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |