Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc34a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080854.3 | NP_543130.2 | 5 - 6 | 542 | 69 | |
GenBank mRNA | AB054999.1 | - | 5 - 6 | 522 | 69 | |
BC125326.1 | - | 4 - 5 | 475 | 40129841 | 69 | |
BC131997.1 | - | 4 - 5 | 475 | 40130274 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |