Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Speer9-ps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_001583.3 | 2 - 3 | 285 | 133 | ||
GenBank mRNA | BY714690.1 | - | 2 - 3 | 286 | 133 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | AK015142.1 | - | 286 | Chr. - - [-] on Build | 133 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Downstream | expressed sequence AU018091 | AU018091 | 18.9kb | Anti-sense | Chr. 7 - 3154658 - 3169252 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |