Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zak | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023057.5 | NP_075544.1 | 16 - 17 | 1611 | 74 | |
GenBank mRNA | AB049731.1 | - | 15 - 16 | 1394 | 74 | |
AK132186.1 | - | 16 - 17 | 1612 | 74 | ||
BC023718.1 | - | 16 - 17 | 1492 | 5346593 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |