Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ola1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025942.2 | NP_080218.1 | 7 - 8 | 888 | 150 | |
GenBank mRNA | AK010817.1 | - | 7 - 8 | 804 | 150 | |
AK013055.1 | - | 7 - 8 | 889 | 150 | ||
AK159873.1 | - | 7 - 8 | 805 | 150 | ||
AK167458.1 | - | 7 - 8 | 804 | 150 | ||
AK170982.1 | - | 7 - 8 | 818 | 150 | ||
BC011318.1 | - | 7 - 8 | 788 | 3985271 | 150 | |
BC029207.1 | - | 7 - 8 | 781 | 4009561 | 150 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |