Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zeb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289521.1 | NP_001276450.1 | 3 - 4 | 587 | 105 | |
NM_015753.4 | NP_056568.2 | 4 - 5 | 924 | 105 | ||
GenBank mRNA | AF033116.1 | - | 3 - 4 | 439 | 105 | |
AK122312.1 | - | 4 - 5 | 497 | 105 | ||
AK141157.1 | - | 3 - 4 | 555 | 105 | ||
BC060699.1 | - | 4 - 5 | 661 | 6408845 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |