Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssfa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080558.4 | NP_542125.3 | 1 - 2 | 324 | 74 | |
GenBank mRNA | AB093303.1 | - | 1 - 2 | 199 | 74 | |
AB120565.1 | - | 1 - 2 | 209 | 74 | ||
AK169543.1 | - | 1 - 2 | 325 | 74 | ||
BC122519.1 | - | 1 - 2 | 210 | 74 | ||
BC141883.1 | - | 1 - 2 | 209 | 40097831 | 74 | |
CF539296.1 | - | 1 - 2 | 140 | 74 | ||
CJ129385.1 | - | 1 - 2 | 373 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |