Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssfa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080558.4 | NP_542125.3 | 3 - 4 | 417 | 70 | |
GenBank mRNA | AB093303.1 | - | 3 - 4 | 292 | 70 | |
AB120565.1 | - | 3 - 4 | 302 | 70 | ||
AK037213.1 | - | 1 - 2 | 127 | 70 | ||
AK169543.1 | - | 3 - 4 | 418 | 70 | ||
BC122519.1 | - | 3 - 4 | 303 | 70 | ||
BC141883.1 | - | 3 - 4 | 302 | 40097831 | 70 | |
CF539296.1 | - | 3 - 4 | 233 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |