Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bdnf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048139.1 | NP_001041604.1 | 1 - 2 | 505 | 152 | |
GenBank mRNA | AY057911.1 | - | 1 - 2 | 505 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Apoptosis Mus musculus Inventoried Probe spans exons |