Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fmn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285458.1 | NP_001272387.1 | 11 - 12 | 3667 | 85 | |
NM_001285459.1 | NP_001272388.1 | 11 - 12 | 3291 | 85 | ||
GenBank mRNA | BC138036.1 | - | 10 - 11 | 3573 | 8861055 | 85 |
BC171945.1 | - | 10 - 11 | 3567 | 9054231 | 85 | |
CK391697.1 | - | 7 - 8 | 593 | 85 | ||
CO814739.2 | - | 6 - 7 | 518 | 85 | ||
X62379.1 | - | 11 - 12 | 3297 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |